missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-3-n-dodecylthiophene, 97%

Manufacturer:  Alfa Aesar™ H55958MD

 View more versions of this product

Catalog No. AAH55958MD



Description

Description

Applications
2,5-Dibromo-3-n-dodecylthiophene is used as an OLED intermediate, flavor and fragrance intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2,5-Dibromo-3-n-dodecylthiophene
>113°C (235°F)
135°C (2.5mmHg)
148256-63-7
MFCD00274315
Slightly soluble in water.
CCCCCCCCCCCCC1=C(SC(=C1)Br)Br
410.252
410.25
1.321
1.53
250mg
C16H26Br2S
2,5-bis bromanyl-3-dodecyl-thiophene
ZIZGNWAMVWNAJT-UHFFFAOYSA-N
2,5-dibromo-3-dodecylthiophene
5072754
97%

Chemical Identifiers

148256-63-7
410.252
ZIZGNWAMVWNAJT-UHFFFAOYSA-N
5072754
CCCCCCCCCCCCC1=C(SC(=C1)Br)Br
C16H26Br2S
MFCD00274315
2,5-bis bromanyl-3-dodecyl-thiophene
2,5-dibromo-3-dodecylthiophene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents