Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-3-n-decylthiophene, 97%

Chemical Identifiers

Chemical Name or Material 2,5-Dibromo-3-n-decylthiophene
CAS 158956-23-1
Formula Weight 382.2
MDL Number MFCD00274314
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5549703

 
alfa aesar™
H5549703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5549706

 
alfa aesar™
H5549706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55497MD

 
alfa aesar™
H55497MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 382.2
Boiling Point 168° to 170°C (2mmHg)
Chemical Name or Material 2,5-Dibromo-3-n-decylthiophene
Physical Form Liquid
Assay 97%
MDL Number MFCD00274314
CAS 158956-23-1
Refractive Index 1.5380
Density 1.372g/mL
Quantity 1g