missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-3-n-decylthiophene, 97%

Manufacturer:  Alfa Aesar™ H5549706

 View more versions of this product

Catalog No. AAH5549706



Specifications

Specifications

2,5-Dibromo-3-n-decylthiophene
113°C (235°F)
168°C to 170°C (2mmHg)
158956-23-1
MFCD00274314
CQDSRZZKSUNTSB-UHFFFAOYSA-N
2,5-dibromo-3-decylthiophene
4218617
Liquid
1.372 g/mL
1.538
5g
C14H22Br2S
2,5-bis bromanyl-3-decyl-thiophene
CCCCCCCCCCC1=C(SC(=C1)Br)Br
382.198
382.2
97%

Chemical Identifiers

158956-23-1
382.198
CQDSRZZKSUNTSB-UHFFFAOYSA-N
4218617
CCCCCCCCCCC1=C(SC(=C1)Br)Br
C14H22Br2S
MFCD00274314
2,5-bis bromanyl-3-decyl-thiophene
2,5-dibromo-3-decylthiophene
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents