missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-3-n-butylthiophene, 96%

$63.34 - $593.28

Chemical Identifiers

CAS 116971-10-9
Molecular Formula C8H10Br2S
Molecular Weight (g/mol) 298.036
MDL Number MFCD00672012
InChI Key YRSQCQUZWSQKKU-UHFFFAOYSA-N
Synonym 2,5-bis bromanyl-3-butyl-thiophene
PubChem CID 600361
IUPAC Name 2,5-dibromo-3-butylthiophene
SMILES CCCCC1=C(SC(=C1)Br)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55465MD View Documents Alfa Aesar™
H55465MD
250mg Each for $63.34
 
AAH5546503 View Documents Alfa Aesar™
H5546503
1g Each for $166.47
 
AAH5546506 View Documents Alfa Aesar™
H5546506
5g Each for $593.28
 
Specifications

Chemical Identifiers

116971-10-9
298.036
YRSQCQUZWSQKKU-UHFFFAOYSA-N
600361
CCCCC1=C(SC(=C1)Br)Br
C8H10Br2S
MFCD00672012
2,5-bis bromanyl-3-butyl-thiophene
2,5-dibromo-3-butylthiophene

Specifications

1.67
1.574
250mg
C8H10Br2S
2,5-bis bromanyl-3-butyl-thiophene
CCCCC1=C(SC(=C1)Br)Br
298.036
298.04
2,5-Dibromo-3-n-butylthiophene
110°C (230°F)
270°C to 281°C
116971-10-9
MFCD00672012
YRSQCQUZWSQKKU-UHFFFAOYSA-N
2,5-dibromo-3-butylthiophene
600361
96%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures