missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-3-n-butylthiophene, 96%

Manufacturer:  Alfa Aesar™ H55465MD

 View more versions of this product

Catalog No. AAH55465MD



Specifications

Specifications

2,5-Dibromo-3-n-butylthiophene
110°C (230°F)
270°C to 281°C
116971-10-9
MFCD00672012
YRSQCQUZWSQKKU-UHFFFAOYSA-N
2,5-dibromo-3-butylthiophene
600361
96%
1.67
1.574
250mg
C8H10Br2S
2,5-bis bromanyl-3-butyl-thiophene
CCCCC1=C(SC(=C1)Br)Br
298.036
298.04

Chemical Identifiers

116971-10-9
298.036
YRSQCQUZWSQKKU-UHFFFAOYSA-N
600361
CCCCC1=C(SC(=C1)Br)Br
C8H10Br2S
MFCD00672012
2,5-bis bromanyl-3-butyl-thiophene
2,5-dibromo-3-butylthiophene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents