Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Diaminotoluene sulfate, 97%

Chemical Identifiers

Chemical Name or Material 2,5-Diaminotoluene sulfate
CAS 615-50-9
Molecular Formula C7H10N2·H2SO4
Formula Weight 220.25
MDL Number MFCD00013003
UN Number UN2811
Beilstein 3755478
Synonym 2-Methyl-1,4-phenylenediamine sulfate; 2,5-Toluenediamine sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0738814

  SDS
alfa aesar™
L0738814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0738822

  SDS
alfa aesar™
L0738822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0738836

  SDS
alfa aesar™
L0738836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.25
Chemical Name or Material 2,5-Diaminotoluene sulfate
Assay 97%
MDL Number MFCD00013003
Health Hazard 2 GHS H Statement
H301-H331-H312-H317
Toxic if swallowed.
Toxic if inhaled.
Harmful in contact with skin.
May cause an allergic skin reaction.
Molecular Formula C7H10N2·H2SO4
CAS 615-50-9
UN Number UN2811
Beilstein 3755478
Synonym 2-Methyl-1,4-phenylenediamine sulfate; 2,5-Toluenediamine sulfate
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
TSCA TSCA
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number XT0525000
Melting Point >300°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H312-H317
Toxic if swallowed.
Toxic if inhaled.
Harmful in contact with skin.
May cause an allergic skin reaction.