missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Diaminotoluene sulfate, 97%

Manufacturer:  Alfa Aesar™ L0738814

 View more versions of this product

Catalog No. AAL0738814



Specifications

Specifications

2,5-Diaminotoluene sulfate
25g
C7H12N2O4S
UN2811
1,4-benzenediamine, 2-methyl-, sulfate
CC1=C(C=CC(=C1)N)N.OS(=O)(=O)O
220.243
220.25
>300°C
615-50-9
MFCD00013003
3755478
KZTWOUOZKZQDMN-UHFFFAOYSA-N
2-methylbenzene-1,4-diamine;sulfuric acid
22856
97%

Chemical Identifiers

615-50-9
220.243
KZTWOUOZKZQDMN-UHFFFAOYSA-N
22856
CC1=C(C=CC(=C1)N)N.OS(=O)(=O)O
C7H12N2O4S
MFCD00013003
1,4-benzenediamine, 2-methyl-, sulfate
2-methylbenzene-1,4-diamine;sulfuric acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H312-H317
Toxic if swallowed.
Toxic if inhaled.
Harmful in contact with skin.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : XT0525000

TSCA : TSCA

Documents