Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Diaminopyridine dihydrochloride, 97%

$59.26 - $59.26

Chemical Identifiers

Chemical Name or Material 2,5-Diaminopyridine dihydrochloride
CAS 26878-35-3
Molecular Formula C5H7N3·2HCl
Formula Weight 182.05
MDL Number MFCD00040818
Beilstein 3912365
Synonym 2,5-Pyridinediamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0559906

 
alfa aesar™
L0559906
5g Each for $59.26
Description & Specifications

Specifications

Formula Weight 182.05
Assay 97%
MDL Number MFCD00040818
CAS 26878-35-3
Beilstein 3912365
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,5-Diaminopyridine dihydrochloride
Molecular Formula C5H7N3·2HCl
Synonym 2,5-Pyridinediamine dihydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈264°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.