Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Diacetoxytoluene, 97%

$50.49 - $161.62

Chemical Identifiers

Chemical Name or Material 2,5-Diacetoxytoluene
CAS 717-27-1
Molecular Formula C11H12O4
Formula Weight 208.22
MDL Number MFCD00061098
Beilstein 1967373
Synonym Methylhydroquinone diacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2315614

 
alfa aesar™
B2315614
25g Each for $50.49

AAB2315622

 
alfa aesar™
B2315622
100g Each for $161.62
Description & Specifications

Specifications

Formula Weight 208.22
Chemical Name or Material 2,5-Diacetoxytoluene
Assay 97%
Molecular Formula C11H12O4
MDL Number MFCD00061098
CAS 717-27-1
Beilstein 1967373
Synonym Methylhydroquinone diacetate
Melting Point 40° to 42°C
Quantity 25g