missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Di-tert-butylhydroquinone, 98+%

Manufacturer:  Alfa Aesar™ A1460630

 View more versions of this product

Catalog No. AAA1460630



Specifications

Specifications

2,5-Di-tert-butylhydroquinone
321°C
250g
C14H22O2
2049542
JZODKRWQWUWGCD-UHFFFAOYSA-N
2,5-ditert-butylbenzene-1,4-diol
2374
222.33
>200°C (392°F)
214°C to 218°C
88-58-4
MFCD00008825
2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
CC(C)(C)C1=CC(=C(C=C1O)C(C)(C)C)O
222.328
CHEBI:41094
≥98%

Chemical Identifiers

88-58-4
222.328
JZODKRWQWUWGCD-UHFFFAOYSA-N
2374
2,5-ditert-butylbenzene-1,4-diol
C14H22O2
MFCD00008825
2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
CHEBI:41094
CC(C)(C)C1=CC(=C(C=C1O)C(C)(C)C)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : MX5160000

TSCA : TSCA

Documents