Please login to your online account to display your discounted pricing

2,5-Di-tert-butyl-p-benzoquinone 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Di-tert-butyl-p-benzoquinone
CAS 2460-77-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14 H20 O2
Linear Formula [(CH3)3C]2C6H2(=O)2
Formula Weight 220.31
MDL Number MFCD00019442
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373661000

  SDS
thermo fisher scientific
AC373661000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC373660250

  SDS
thermo fisher scientific
AC373660250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC373660050

  SDS
thermo fisher scientific
AC373660050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 220.31
Physical Form Crystalline Powder
Quantity 100g
Color Yellow to golden
Melting Point 152° to 154°C
CAS Max % 100.0
Assay Percent Range 99%
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00019442
CAS Min % 98.5
Packaging Glass bottle
Linear Formula [(CH3)3C]2C6H2(=O)2
Chemical Name or Material 2, 5-Di-tert-butyl-p-benzoquinone
Molecular Formula C14 H20 O2
CAS 2460-77-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.