Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Di-tert-butyl-p-benzoquinone, 99%

$42.95 - $42.95

Chemical Identifiers

Chemical Name or Material 2,5-Di-tert-butyl-p-benzoquinone
CAS 2460-77-7
Molecular Formula C14H20O2
Formula Weight 220.31
MDL Number MFCD00019442
Beilstein 2047945
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1005406

  SDS
alfa aesar™
L1005406
5g Each for $42.95
Description & Specifications

Specifications

Formula Weight 220.31
MDL Number MFCD00019442
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 2460-77-7
Chemical Name or Material 2,5-Di-tert-butyl-p-benzoquinone
Health Hazard 1 Warning
Molecular Formula C14H20O2
Quantity 5g
Assay 99%
Melting Point 152° to 153°C
RTECS Number GU5140000
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2047945
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.