Please login to your online account to display your discounted pricing

2,5-Di(2,2,2-trifluoroethoxy)benzene-1-carbohydrazide, ≥95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB03377EA

 View more versions of this product

Catalog No. SPB03377EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay 90%
Formula Weight 332.2
Quantity 10g
CAS Min % 90.0
Assay Percent Range 90%+
Molecular Formula C11H10F6N2O3
MDL Number MFCD00221192
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2,5-Di(2,2,2-trifluoroethoxy)benzene-1-carbohydrazide
Packaging Amber Glass Bottle
CAS 50778-75-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2,5-Di(2,2,2-trifluoroethoxy)benzene-1-carbohydrazide
CAS 50778-75-1
CAS Min % 90.0
Assay 90%
Assay Percent Range 90%+
Molecular Formula C11H10F6N2O3
Formula Weight 332.2
MDL Number MFCD00221192
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.