Please login to your online account to display your discounted pricing

2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid
CAS 17266-30-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7ClO2S
Formula Weight 226.68
MDL Number MFCD00052308
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MWP00086CB

  SDS
fisher scientific company
MWP00086CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.68
Quantity 250mg
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C10H7ClO2S
Chemical Name or Material 2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid
MDL Number MFCD00052308
CAS 17266-30-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.