Please login to your online account to display your discounted pricing

2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MWP00086CB

 View more versions of this product

Catalog No. MWP00086CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 226.68
Quantity 250mg
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C10H7ClO2S
Chemical Name or Material 2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid
MDL Number MFCD00052308
CAS 17266-30-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid
CAS 17266-30-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7ClO2S
Formula Weight 226.68
MDL Number MFCD00052308
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents