Please login to your online account to display your discounted pricing

2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MWP00226DA

 View more versions of this product

Catalog No. MWP00226DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 240.71
Quantity 1g
Assay 95%
CAS Min % 95.0
CAS 51527-19-6
Chemical Name or Material 2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid
Assay Percent Range 95%
Molecular Formula C11H9ClO2S
MDL Number MFCD00052300
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material 2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid
CAS 51527-19-6
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H9ClO2S
Formula Weight 240.71
MDL Number MFCD00052300
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.