missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol, TCI America™

$82.50 - $261.25

Chemical Identifiers

CAS 3896-11-5
Molecular Formula C17H18ClN3O
Molecular Weight (g/mol) 315.80
MDL Number MFCD00059707
InChI Key OCWYEMOEOGEQAN-UHFFFAOYSA-N
Synonym 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chlorobenzotriazole
PubChem CID 62531
IUPAC Name 2-tert-butyl-6-(5-chlorobenzotriazol-2-yl)-4-methylphenol
SMILES CC1=CC(=C(C(=C1)N2N=C3C=CC(=CC3=N2)Cl)O)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H055925G View Documents TCI America
H055925G
25g Each for $82.50
 
H0559500G View Documents TCI America
H0559500G
500g Each for $261.25
 
Specifications

Chemical Identifiers

3896-11-5
315.80
OCWYEMOEOGEQAN-UHFFFAOYSA-N
62531
CC1=CC(=C(C(=C1)N2N=C3C=CC(=CC3=N2)Cl)O)C(C)(C)C
C17H18ClN3O
MFCD00059707
2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chlorobenzotriazole
2-tert-butyl-6-(5-chlorobenzotriazol-2-yl)-4-methylphenol

Specifications

Yellow
25g
C17H18ClN3O
2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chlorobenzotriazole
CC1=CC(=C(C(=C1)N2N=C3C=CC(=CC3=N2)Cl)O)C(C)(C)C
315.80
315.80
≥98.0% (GC)
140°C
3896-11-5
MFCD00059707
OCWYEMOEOGEQAN-UHFFFAOYSA-N
2-tert-butyl-6-(5-chlorobenzotriazol-2-yl)-4-methylphenol
62531
Crystal-Powder at 20°C
2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0545

TSCA : Yes