missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(5-Bromo-2-pyridylazo)-5-dimethylaminophenol [for Colorimetric Determination of Cd], TCI America™

$310.14 - $1,695.88

Chemical Identifiers

CAS 50783-82-9
Molecular Formula C13H13BrN4O
Molecular Weight (g/mol) 321.178
MDL Number MFCD00059779
InChI Key GXYVDXVMTIAMFF-WJDWOHSUSA-N
Synonym 2-(4-Dimethylamino-2-hydroxyphenylazo)-5-bromopyridine
PubChem CID 22323640
IUPAC Name (6Z)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(dimethylamino)cyclohexa-2,4-dien-1-one
SMILES CN(C)C1=CC(=O)C(=NNC2=NC=C(C=C2)Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B1082100MG View Documents TCI America
B1082100MG
100mg Each for $310.14
 
B10821G View Documents TCI America
B10821G
1g Each for $1,695.88
 
Specifications

Chemical Identifiers

50783-82-9
321.178
GXYVDXVMTIAMFF-WJDWOHSUSA-N
22323640
CN(C)C1=CC(=O)C(=NNC2=NC=C(C=C2)Br)C=C1
C13H13BrN4O
MFCD00059779
2-(4-Dimethylamino-2-hydroxyphenylazo)-5-bromopyridine
(6Z)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(dimethylamino)cyclohexa-2,4-dien-1-one

Specifications

Purple-Red
100mg
C13H13BrN4O
2-(4-Dimethylamino-2-hydroxyphenylazo)-5-bromopyridine
CN(C)C1=CC(=O)C(=NNC2=NC=C(C=C2)Br)C=C1
321.178
321.18
≥93.0% (HPLC)
217°C
50783-82-9
MFCD00059779
GXYVDXVMTIAMFF-WJDWOHSUSA-N
(6Z)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(dimethylamino)cyclohexa-2,4-dien-1-one
22323640
Crystal-Powder at 20°C
2-(5-Bromo-2-pyridylazo)-5-dimethylaminophenol [for Colorimetric Determination of Cd]
Safety and Handling

Safety and Handling

TSCA : No