Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(5-Bromo-2-methoxyphenyl)ethylamine hydrobromide, 98%

$394.58 - $394.58

Chemical Identifiers

Chemical Name or Material 2-(5-Bromo-2-methoxyphenyl)ethylamine hydrobromide
CAS 206559-44-6
Molecular Formula C9H12BrNO·HBr
Formula Weight 311.02g/mol
MDL Number MFCD00238752
Synonym 5-Bromo-2-methoxyphenethylamine hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1987506

 
alfa aesar™
L19875-06
5g Each for $394.58
Description & Specifications

Specifications

Quantity 5g
Melting Point 202° to 204°C
Quantity 5g
Chemical Name or Material 2-(5-Bromo-2-methoxyphenyl)ethylamine hydrobromide
CAS 206559-44-6
Assay 98%
Molecular Formula C9H12BrNO·HBr
Formula Weight 311.02g/mol
MDL Number MFCD00238752
Synonym 5-Bromo-2-methoxyphenethylamine hydrobromide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.