Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(trimethylsilyl)thiazole, 96%

$407.93 - $407.93

Chemical Identifiers

Chemical Name or Material 2,5-Bis(trimethylsilyl)thiazole
CAS 79265-34-2
Molecular Formula C9H19NSSi2
Formula Weight 229.49g/mol
MDL Number MFCD09800395
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2922206

 
alfa aesar™
H29222-06
5g Each for $407.93
Description & Specifications

Specifications

Melting Point 53° to 57°C
Boiling Point 100°C (13mmHg)
Quantity 5g
Chemical Name or Material 2,5-Bis(trimethylsilyl)thiazole
CAS 79265-34-2
Assay 96%
Molecular Formula C9H19NSSi2
Formula Weight 229.49g/mol
MDL Number MFCD09800395
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C