Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2,5-Bis(trifluoromethyl)benzenesulfonyl chloride
CAS 351003-22-0
Molecular Formula C8H3ClF6O2S
Formula Weight 312.61
MDL Number MFCD03094042
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1987803

 
alfa aesar™
L1987803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1987806

 
alfa aesar™
L1987806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 312.61
Chemical Name or Material 2,5-Bis(trifluoromethyl)benzenesulfonyl chloride
CAS 351003-22-0
UN Number UN3261
Melting Point 59° to 63°C
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 97%
Molecular Formula C8H3ClF6O2S
MDL Number MFCD03094042
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.