Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Bis(trifluoromethyl)acetophenone, 97%

$91.46 - $506.27

Chemical Identifiers

Chemical Name or Material 2',5'-Bis(trifluoromethyl)acetophenone
CAS 545410-47-7
Molecular Formula C10H6F6O
Formula Weight 256.15
MDL Number MFCD04116044
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2665903

  SDS
alfa aesar™
H2665903
1g Each for $91.46

AAH2665909

  SDS
alfa aesar™
H2665909
10g Each for $506.27
Description & Specifications

Specifications

Formula Weight 256.15
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD04116044
Chemical Name or Material 2',5'-Bis(trifluoromethyl)acetophenone
CAS 545410-47-7
Molecular Formula C10H6F6O
Health Hazard 1 Warning
Quantity 1g

Applications
Bis(trifluoromethyl)acetophenone is used in the synthesis of the monoamine oxidase-B inhibitors. It is also used to isolate novel bacterial strain from soil samples.

Solubility
Sparingly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.