Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(diethylamino)phenyl-1,3,4-oxadiazole, 98%

$177.01 - $177.01

Chemical Identifiers

Chemical Name or Material 2,5-Bis(diethylamino)phenyl-1,3,4-oxadiazole
CAS 1679-98-7
Molecular Formula C22H28N4O
Formula Weight 364.49
MDL Number MFCD00798595
Beilstein 569499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2090122

 
alfa aesar™
B2090122
100g Each for $177.01
Description & Specifications

Specifications

Formula Weight 364.49
Chemical Name or Material 2,5-Bis(diethylamino)phenyl-1,3,4-oxadiazole
TSCA TSCA
Assay 98%
Molecular Formula C22H28N4O
MDL Number MFCD00798595
CAS 1679-98-7
Beilstein 569499
Melting Point 148° to 151°C
Quantity 100g