Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(chloromethyl)-p-xylene, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Bis(chloromethyl)-p-xylene
CAS 6298-72-2
Molecular Formula C10H12Cl2
Formula Weight 203.11g/mol
MDL Number MFCD00013682
UN Number UN1759
Synonym alpha(1),alpha(4)-Dichlorodurene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1314706

 
alfa aesar™
A13147-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1314714

 
alfa aesar™
A13147-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1314722

 
alfa aesar™
A13147-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 130° to 133°C
Quantity 5g
Chemical Name or Material 2,5-Bis(chloromethyl)-p-xylene
CAS 6298-72-2
Assay 98%
Molecular Formula C10H12Cl2
Formula Weight 203.11g/mol
MDL Number MFCD00013682
UN Number UN1759
Synonym alpha(1),alpha(4)-Dichlorodurene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
RTECS Number CY8424500
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.