Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole
CAS 146370-52-7
Molecular Formula C17H26Cl2O2
Formula Weight 333.29
MDL Number MFCD03093939
UN Number UN1759
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5104103

 
alfa aesar™
H5104103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5104106

 
alfa aesar™
H5104106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 333.29
Chemical Name or Material 2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole
Health Hazard 1 Danger
Assay 98%
Molecular Formula C17H26Cl2O2
MDL Number MFCD03093939
CAS 146370-52-7
UN Number UN1759
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

Applications
2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole is used for conducting polymer precursor. It is also used as organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.