Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(4-nitrophenyl)-1,3,4-oxadiazole, 97%

$88.59 - $88.59

Chemical Identifiers

Chemical Name or Material 2,5-Bis(4-nitrophenyl)-1,3,4-oxadiazole
CAS 1044-49-1
Molecular Formula C14H8N4O5
Formula Weight 312.24
MDL Number MFCD00173659
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2096106

  SDS
alfa aesar™
B2096106
5g Each for $88.59
Description & Specifications

Specifications

Formula Weight 312.24
CAS 1044-49-1
Melting Point 310° to 312°C
RTECS Number RO0674000
Chemical Name or Material 2,5-Bis(4-nitrophenyl)-1,3,4-oxadiazole
Assay 97%
Quantity 5g
MDL Number MFCD00173659
Molecular Formula C14H8N4O5