Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(4-biphenylyl)oxazole, 99%

$122.05 - $2,333.97

Chemical Identifiers

Chemical Name or Material 2,5-Bis(4-biphenylyl)oxazole
CAS 2083-09-2
Molecular Formula C27H19NO
Formula Weight 373.46
MDL Number MFCD00005305
Beilstein 305439
Synonym BBO; 2,5-Di(4-biphenylyl)oxazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1362303

  SDS
alfa aesar™
A1362303
1g Each for $122.05

AAA1362314

  SDS
alfa aesar™
A1362314
25g Each for $2,333.97

AAA1362306

  SDS
alfa aesar™
A1362306
5g Each for $531.89
Description & Specifications

Specifications

Formula Weight 373.46
MDL Number MFCD00005305
Chemical Name or Material 2,5-Bis(4-biphenylyl)oxazole
Melting Point 238° to 240°C
Assay 99%
Molecular Formula C27H19NO
Beilstein 305439
Quantity 1g
Synonym BBO; 2,5-Di(4-biphenylyl)oxazole
CAS 2083-09-2