missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid, 96%

Manufacturer:  Alfa Aesar™ H6110322

 View more versions of this product

Catalog No. AAH6110322



Specifications

Specifications

2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid
100g
C11H8F6O4
2,5-bis 2,2,2-trifluoroethoxy benzoic acid, 2,5-bis trifluoroethoxy benzoic acid, 2,5-di 2,2,2-trifluoroethoxy benzoic acid, benzoic acid, 2,5-bis 2,2,2-trifluoroethoxy, flecainide impurity d, pubchem10082, acmc-209ih5, 2,5-di 2,2,2-trifluoroethoxy benzoicacid, 2,5-bis 2,2,2-trifluorethoxy benzoic acid, 2,5-bis 2,2,2-trifluoroethoxy benzoicacid
C1=CC(=C(C=C1OCC(F)(F)F)C(=O)O)OCC(F)(F)F
318.171
318.17
96%
White
35480-52-5
MFCD00221424
YPGYLCZBZKRYQJ-UHFFFAOYSA-N
2,5-bis(2,2,2-trifluoroethoxy)benzoic acid
2736078
Solid

Chemical Identifiers

35480-52-5
318.171
YPGYLCZBZKRYQJ-UHFFFAOYSA-N
2736078
C1=CC(=C(C=C1OCC(F)(F)F)C(=O)O)OCC(F)(F)F
C11H8F6O4
MFCD00221424
2,5-bis 2,2,2-trifluoroethoxy benzoic acid, 2,5-bis trifluoroethoxy benzoic acid, 2,5-di 2,2,2-trifluoroethoxy benzoic acid, benzoic acid, 2,5-bis 2,2,2-trifluoroethoxy, flecainide impurity d, pubchem10082, acmc-209ih5, 2,5-di 2,2,2-trifluoroethoxy benzoicacid, 2,5-bis 2,2,2-trifluorethoxy benzoic acid, 2,5-bis 2,2,2-trifluoroethoxy benzoicacid
2,5-bis(2,2,2-trifluoroethoxy)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents