Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Bis(1-naphthyl)-1,3,4-oxadiazole, 99%

$204.92 - $204.92

Chemical Identifiers

Chemical Name or Material 2,5-Bis(1-naphthyl)-1,3,4-oxadiazole
CAS 905-62-4
Molecular Formula C22H14N2O
Formula Weight 322.37
MDL Number MFCD00045582
Beilstein 285250
Synonym 2,5-Di(1-naphthyl)-1,3,4-oxadiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0721506

 
alfa aesar™
L0721506
5g Each for $204.92
Description & Specifications

Specifications

Formula Weight 322.37
Chemical Name or Material 2,5-Bis(1-naphthyl)-1,3,4-oxadiazole
MDL Number MFCD00045582
CAS 905-62-4
Synonym 2,5-Di(1-naphthyl)-1,3,4-oxadiazole
Molecular Formula C22H14N2O
Beilstein 285250
Assay 99%
Melting Point 175° to 178°C
Quantity 5g