missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone, TCI America™

$171.11 - $1,459.06

Chemical Identifiers

CAS 903-19-5
Molecular Formula C22H38O2
Molecular Weight (g/mol) 334.544
MDL Number MFCD00128797
InChI Key CLDZVCMRASJQFO-UHFFFAOYSA-N
PubChem CID 70180
IUPAC Name 2,5-bis(2,4,4-trimethylpentan-2-yl)benzene-1,4-diol
SMILES CC(C)(C)CC(C)(C)C1=CC(=C(C=C1O)C(C)(C)CC(C)(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B183925G View Documents TCI America
B183925G
25g Each for $171.11
 
B1839500G View Documents TCI America
B1839500G
500g Each for $1,459.06
 
Specifications

Chemical Identifiers

903-19-5
334.544
CLDZVCMRASJQFO-UHFFFAOYSA-N
2,5-bis(2,4,4-trimethylpentan-2-yl)benzene-1,4-diol
C22H38O2
MFCD00128797
70180
CC(C)(C)CC(C)(C)C1=CC(=C(C=C1O)C(C)(C)CC(C)(C)C)O

Specifications

Undesignated
25g
C22H38O2
CLDZVCMRASJQFO-UHFFFAOYSA-N
2,5-bis(2,4,4-trimethylpentan-2-yl)benzene-1,4-diol
70180
Crystal-Powder at 20°C
2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone
130°C
903-19-5
MFCD00128797
CC(C)(C)CC(C)(C)C1=CC(=C(C=C1O)C(C)(C)CC(C)(C)C)O
334.544
334.54
≥95.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0553

TSCA : Yes