Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5,6-Trimethylbenzoxazole, 98%

$33.59 - $441.49

Chemical Identifiers

Chemical Name or Material 2,5,6-Trimethylbenzoxazole
CAS 19219-98-8
Molecular Formula C10H11NO
Formula Weight 161.2
MDL Number MFCD00022849
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2261703

  SDS
alfa aesar™
B2261703
1g Each for $33.59

AAB2261706

  SDS
alfa aesar™
B2261706
5g Each for $116.03

AAB2261714

  SDS
alfa aesar™
B2261714
25g Each for $441.49
Description & Specifications

Specifications

Formula Weight 161.2
Melting Point 91° to 96°C
MDL Number MFCD00022849
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 19219-98-8
Molecular Formula C10H11NO
Assay 98%
Quantity 1g
Chemical Name or Material 2,5,6-Trimethylbenzoxazole
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.