Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5,6-Trimethylbenzothiazole, 98%

$45.05 - $323.96

Chemical Identifiers

Chemical Name or Material 2,5,6-Trimethylbenzothiazole
CAS 5683-41-0
Molecular Formula C10H11NS
Formula Weight 177.26
MDL Number MFCD00128832
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2184603

  SDS
alfa aesar™
B2184603
1g Each for $45.05

AAB2184614

  SDS
alfa aesar™
B2184614
25g Each for $323.96

AAB2184606

  SDS
alfa aesar™
B2184606
5g Each for $138.47
Description & Specifications

Specifications

Formula Weight 177.26
MDL Number MFCD00128832
Molecular Formula C10H11NS
Boiling Point 143°C (5mmHg)
Melting Point 83° to 85°C
Quantity 1g
Assay 98%
Odor Odorless
CAS 5683-41-0
Chemical Name or Material 2,5,6-Trimethylbenzothiazole