Learn More
2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6441506
Description
It is applied as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied as an active pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
2-[4-(Trifluoromethyl)phenyl]ethylamine | |
C9H10F3N | |
5 g | |
2719818 | |
Slightly soluble in water. | |
C1=CC(=CC=C1CCN)C(F)(F)F | |
189.181 | |
189.18 |
775-00-8 | |
MFCD01529875 | |
UN2735 | |
4-trifluoromethylphenethylamine, 2-4-trifluoromethyl-phenyl-ethylamine, 2-4-trifluoromethyl phenyl ethanamine, 4-trifluoromethyl phenethylamine, 2-4-trifluoromethyl-phenyl ethylamine, 2-4-trifluoromethylphenyl ethylamine, 2-4-trifluoromethyl phenyl ethan-1-amine, 2-4-trifluoromethyl phenyl ethylamine, 2-4-trifluoromethyl phenyl ethyl amine | |
HMFOBPNVAAAACP-UHFFFAOYSA-N | |
2-[4-(trifluoromethyl)phenyl]ethanamine | |
1988106 | |
98% |
Chemical Identifiers
775-00-8 | |
189.181 | |
HMFOBPNVAAAACP-UHFFFAOYSA-N | |
1988106 | |
C1=CC(=CC=C1CCN)C(F)(F)F |
C9H10F3N | |
MFCD01529875 | |
4-trifluoromethylphenethylamine, 2-4-trifluoromethyl-phenyl-ethylamine, 2-4-trifluoromethyl phenyl ethanamine, 4-trifluoromethyl phenethylamine, 2-4-trifluoromethyl-phenyl ethylamine, 2-4-trifluoromethylphenyl ethylamine, 2-4-trifluoromethyl phenyl ethan-1-amine, 2-4-trifluoromethyl phenyl ethylamine, 2-4-trifluoromethyl phenyl ethyl amine | |
2-[4-(trifluoromethyl)phenyl]ethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only