Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Trifluoromethyl-alpha-toluenesulfonyl)thioacetamide, 97%

$69.58 - $69.58

Chemical Identifiers

Chemical Name or Material 2-(4-Trifluoromethyl-alpha-toluenesulfonyl)thioacetamide
Molecular Formula C10H10F3NO2S2
Formula Weight 297.32g/mol
MDL Number MFCD09757596
Synonym 2-[4-(Trifluoromethyl)benzylsulfonyl]thioacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2626403

 
alfa aesar™
H26264-03
1g Each for $69.58
Description & Specifications

Specifications

Melting Point 90° to 92°C
Quantity 1g
Chemical Name or Material 2-(4-Trifluoromethyl-alpha-toluenesulfonyl)thioacetamide
Assay 97%
Molecular Formula C10H10F3NO2S2
Formula Weight 297.32g/mol
MDL Number MFCD09757596
Synonym 2-[4-(Trifluoromethyl)benzylsulfonyl]thioacetamide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.