Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Toluoyl)benzoic acid, 98%

$37.36 - $614.76

Chemical Identifiers

Chemical Name or Material 2-(4-Toluoyl)benzoic acid
CAS 85-55-2
Molecular Formula C15H12O3
Formula Weight 240.26
MDL Number MFCD00020287
Beilstein 2111078
Merck Index 14,9539
Synonym 2-(4-Methylbenzoyl)benzoic acid; 4'-Methylbenzophenone-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA127950E

  SDS
alfa aesar™
A127950E
2500g Each for $614.76

AAA1279522

  SDS
alfa aesar™
A1279522
100g Each for $37.36

AAA1279536

  SDS
alfa aesar™
A1279536
500g Each for $140.74
Description & Specifications

Specifications

Formula Weight 240.26
Quantity 2500g
CAS 85-55-2
Melting Point 137° to 139°C
TSCA TSCA
MDL Number MFCD00020287
Molecular Formula C15H12O3
Merck Index 14,9539
Chemical Name or Material 2-(4-Toluoyl)benzoic acid
Synonym 2-(4-Methylbenzoyl)benzoic acid; 4'-Methylbenzophenone-2-carboxylic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Beilstein 2111078
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.