Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Thiocarbamoylphenyl)benzimidazole, 97%

Chemical Identifiers

Chemical Name or Material 2-(4-Thiocarbamoylphenyl)benzimidazole
Molecular Formula C14H11N3S
Formula Weight 253.32
MDL Number MFCD16251568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5179203

  SDS
alfa aesar™
H5179203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH51792MD

  SDS
alfa aesar™
H51792MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 253.32
MDL Number MFCD16251568
Quantity 1g
Chemical Name or Material 2-(4-Thiocarbamoylphenyl)benzimidazole
Health Hazard 3 GHS P Statement
P280f-P305+P351+P338
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C14H11N3S
Safety and Handling

Warning

GHS P Statement
P280f-P305+P351+P338
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.