missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Thiazolidinedione, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC159495000

 View more versions of this product

Catalog No. AC159495000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4-Thiazolidinedione
Authentic
178°C to 179°C (19.0mmHg)
123°C to 126°C
2295-31-0
100.0
C3H3NO2S
27, 233
ZOBPZXTWZATXDG-UHFFFAOYSA-N
1,3-thiazolidine-2,4-dione
5437
117.12
99%
138°C
Glass bottle
Yellow
500g
98.5
98.5% min. (HPLC)
MFCD00005478
2,4 3h,5h-thiazoledione
C1C(=O)NC(=O)S1
117.12
CHEBI:50992
Crystalline Powder

Chemical Identifiers

2295-31-0
117.12
ZOBPZXTWZATXDG-UHFFFAOYSA-N
5437
1,3-thiazolidine-2,4-dione
C3H3NO2S
MFCD00005478
2,4 3h,5h-thiazoledione
CHEBI:50992
C1C(=O)NC(=O)S1
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 218-941-2

SDS Documents