Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Thiazolidinedione, 99%

$236.57 - $236.57

Chemical Identifiers

Chemical Name or Material 2,4-Thiazolidinedione
CAS 2295-31-0
Molecular Formula C3H3NO2S
Formula Weight 117.13
MDL Number MFCD00005478
Beilstein 110700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1349622

  SDS
alfa aesar™
A1349622
100g Each for $236.57
Description & Specifications

Specifications

Formula Weight 117.13
Chemical Name or Material 2,4-Thiazolidinedione
TSCA TSCA
Assay 99%
Molecular Formula C3H3NO2S
MDL Number MFCD00005478
CAS 2295-31-0
Beilstein 110700
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XJ5775000
Boiling Point 178° to 179°C (19mmHg)
Melting Point 123° to 126°C
Quantity 100g

Applications
2,4-Thiazolidinedione may be used in the synthesis of sixteen 5-arylidene-2,4-thiazolidinediones, via ultrasound-promoted aldol condensation reaction. It may be used as starting material for the synthesis of drugs with antihyperglycemic activity.

Solubility
Soluble in water (30 mg/ml at 3°C), methanol, DMSO, ethanol, and ethyl ether.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.