Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester, 95%

$87.28 - $785.03

Chemical Identifiers

Chemical Name or Material 2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester
Molecular Formula C18H29BO2
Formula Weight 288.24
MDL Number MFCD09953477
Synonym 2-[2-(4-tert-Butylphenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2705103

 
alfa aesar™
H2705103
1g Each for $87.28

AAH2705106

 
alfa aesar™
H2705106
5g Each for $257.05

AAH2705114

 
alfa aesar™
H2705114
25g Each for $785.03
Description & Specifications

Specifications

Formula Weight 288.24
Chemical Name or Material 2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester
Assay 95%
Molecular Formula C18H29BO2
MDL Number MFCD09953477
Synonym 2-[2-(4-tert-Butylphenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Quantity 1g