Learn More
2-(4-tert-Butylphenyl)ethanol, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1361114
Description
2-(4-tert-Butylphenyl)ethanol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(4-tert-Butylphenyl)ethanol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
2-(4-tert-Butylphenyl)ethanol | |
31°C to 35°C | |
141°C to 143°C (15 mmHg) | |
1.5209 | |
25 g | |
2246595 | |
Insoluble in water. | |
CC(C)(C)C1=CC=C(C=C1)CCO | |
178.275 | |
178.28 |
5406-86-0 | |
0.975 | |
C12H18O | |
MFCD00236023 | |
UN3077 | |
2-4-tert-butylphenyl ethanol, 2-4-tert-butyl phenyl ethanol, p-tert-butylphenethyl alcohol, p-tert-butyl phenethyl alcohol, phenethyl alcohol, p-tert-butyl, 4-tert-butylphenethyl alcohol, unii-aes74i7nru, aes74i7nru, benzeneethanol, 4-1,1-dimethylethyl | |
NZGMMENPUKHODD-UHFFFAOYSA-N | |
2-(4-tert-butylphenyl)ethanol | |
79410 | |
96% |
Chemical Identifiers
5406-86-0 | |
178.275 | |
NZGMMENPUKHODD-UHFFFAOYSA-N | |
79410 | |
CC(C)(C)C1=CC=C(C=C1)CCO |
C12H18O | |
MFCD00236023 | |
2-4-tert-butylphenyl ethanol, 2-4-tert-butyl phenyl ethanol, p-tert-butylphenethyl alcohol, p-tert-butyl phenethyl alcohol, phenethyl alcohol, p-tert-butyl, 4-tert-butylphenethyl alcohol, unii-aes74i7nru, aes74i7nru, benzeneethanol, 4-1,1-dimethylethyl | |
2-(4-tert-butylphenyl)ethanol |
Safety and Handling
GHS H Statement
H318-H361-H373
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c
H318-H361f-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 410-020-5
RTECSNumber : SG7875000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only