missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Quinolinediol, 97%, ACROS Organics™

Chemical Identifiers

CAS 86-95-3
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
InChI Key HDHQZCHIXUUSMK-UHFFFAOYSA-N
Synonym 2 1h-quinolinone, 4-hydroxy
PubChem CID 54680871
ChEBI CHEBI:75926
IUPAC Name 4-hydroxy-1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC132200100 SDS View Documents Thermo Fisher Scientific
132200100
10g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

86-95-3
161.16
2 1h-quinolinone, 4-hydroxy
CHEBI:75926
C1=CC=C2C(=C1)C(=CC(=O)N2)O
C9H7NO2
HDHQZCHIXUUSMK-UHFFFAOYSA-N
54680871
4-hydroxy-1H-quinolin-2-one

Specifications

Authentic
Brown
86-95-3
100.0
C9H7NO2
2 1h-quinolinone, 4-hydroxy
C1=CC=C2C(=C1)C(=CC(=O)N2)O
161.16
CHEBI:75926
Powder
2, 4-Quinolinediol
Glass bottle
10g
96.0
97%
21, 171
HDHQZCHIXUUSMK-UHFFFAOYSA-N
4-hydroxy-1H-quinolin-2-one
54680871
161.16
≥96.0%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning