missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Pyridyl)imidazole-4,5-dithiocarboxamide, 97%

Manufacturer:  Alfa Aesar™ H54733MD

 View more versions of this product

Catalog No. AAH54733MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(4-Pyridyl)imidazole-4,5-dithiocarboxamide
1347815-03-5
MFCD20265330
DZNBSARSJBFIQR-UHFFFAOYSA-N
2-pyridin-4-yl-1H-imidazole-4,5-dicarbothioamide
73995943
97%
250mg
C10H9N5S2
2-4-pyridyl imidazole-4,5-dithiocarboxamide
C1=CN=CC=C1C2=NC(=C(N2)C(=S)N)C(=S)N
263.337
263.33

Chemical Identifiers

1347815-03-5
263.337
DZNBSARSJBFIQR-UHFFFAOYSA-N
73995943
C1=CN=CC=C1C2=NC(=C(N2)C(=S)N)C(=S)N
C10H9N5S2
MFCD20265330
2-4-pyridyl imidazole-4,5-dithiocarboxamide
2-pyridin-4-yl-1H-imidazole-4,5-dicarbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents