missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5492006

 View more versions of this product

Catalog No. AAH5492006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(4-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione
1347815-30-8
MFCD20265373
YZNTXTFZKAVVRN-UHFFFAOYSA-N
2-pyridin-4-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
73996030
97%
5g
C20H11N3O2
2-pyridin-4-yl-1h-anthra 1,2-d imidazole-6,11-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CC=NC=C5
325.327
325.33

Chemical Identifiers

1347815-30-8
325.327
YZNTXTFZKAVVRN-UHFFFAOYSA-N
73996030
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CC=NC=C5
C20H11N3O2
MFCD20265373
2-pyridin-4-yl-1h-anthra 1,2-d imidazole-6,11-dione
2-pyridin-4-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents