Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Piperidinyloxy)-N-propylacetamide, 95%

$93.27 - $337.51

Chemical Identifiers

Chemical Name or Material 2-(4-Piperidinyloxy)-N-propylacetamide
CAS 902836-22-0
Molecular Formula C10H20N2O2
Formula Weight 200.28
MDL Number MFCD08064265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5086503

  SDS
alfa aesar™
H5086503
1g Each for $337.51

AAH50865MD

  SDS
alfa aesar™
H50865MD
250mg Each for $93.27
Description & Specifications

Specifications

Formula Weight 200.28
Assay 95%
Molecular Formula C10H20N2O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(4-Piperidinyloxy)-N-propylacetamide
CAS 902836-22-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Health Hazard 1 Warning
MDL Number MFCD08064265
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.