Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Pentanedione dioxime, 98+%

$67.58 - $755.78

Chemical Identifiers

Chemical Name or Material 2,4-Pentanedione dioxime
CAS 2157-56-4
Molecular Formula C5H10N2O2
Formula Weight 130.15
MDL Number MFCD00013932
Beilstein 4372703
Synonym Acetylacetone dioxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1300306

 
alfa aesar™
A1300306
5g Each for $67.58

AAA1300314

 
alfa aesar™
A1300314
25g Each for $238.59

AAA1300322

 
alfa aesar™
A1300322
100g Each for $755.78
Description & Specifications

Specifications

Formula Weight 130.15
Chemical Name or Material 2,4-Pentanedione dioxime
TSCA TSCA
Assay ≥98%
Molecular Formula C5H10N2O2
MDL Number MFCD00013932
CAS 2157-56-4
Beilstein 4372703
Synonym Acetylacetone dioxime
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 147° to 151°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.