missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2,4-Pentanedionato)lithium, TCI America™

Manufacturer:  TCI America L0080500G

 View more versions of this product

Catalog No. L0080500G



Specifications

Specifications

(2,4-Pentanedionato)lithium
250°C
18115-70-3
MFCD00013496
106.05
≥98.0% (T)
White-Yellow
500g
C5H7LiO2
Acetylacetone Lithium Salt, Lithium Acetylacetonate
Crystal-Powder at 20°C

Chemical Identifiers

18115-70-3
C5H7LiO2
MFCD00013496
Acetylacetone Lithium Salt, Lithium Acetylacetonate
Safety and Handling

Safety and Handling

TSCA : No

Documents