missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2,4-Pentanedionato)lithium, TCI America™

$47.36 - $470.56

Chemical Identifiers

CAS 18115-70-3
Molecular Formula C5H7LiO2
MDL Number MFCD00013496
Synonym Acetylacetone Lithium Salt, Lithium Acetylacetonate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L008025G View Documents TCI America
L008025G
25g Each for $47.36
 
L0080500G View Documents TCI America
L0080500G
500g Each for $470.56
 
Specifications

Chemical Identifiers

18115-70-3
C5H7LiO2
MFCD00013496
Acetylacetone Lithium Salt, Lithium Acetylacetonate

Specifications

White-Yellow
25g
C5H7LiO2
Acetylacetone Lithium Salt, Lithium Acetylacetonate
Crystal-Powder at 20°C
(2,4-Pentanedionato)lithium
250°C
18115-70-3
MFCD00013496
106.05
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No