missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Pentanediol, (+/-) + meso, 99%

$108.84 - $361.16

Chemical Identifiers

CAS 625-69-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00004549
InChI Key GTCCGKPBSJZVRZ-UHFFFAOYSA-N
Synonym 2,4-amylene glycol, 2,4-amyleneglycol, 2,4-dihydroxypentane, 2,4-pentandiol, 2,4-pentanediol, 2,4-pentanediol dl+meso, dl-2,4-pentanediol, isoamylene alcohol, pentanediol-2,4, wln: qy&1yq
PubChem CID 12262
IUPAC Name pentane-2,4-diol
SMILES CC(CC(C)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0177909
View Documents
Alfa Aesar™
L0177909
10g Each for $108.84
 
AAL0177918
View Documents
Alfa Aesar™
L0177918
50g Each for $361.16
 
Description

Description

Applications
2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.

Solubility
Fully miscible.

Notes
Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

625-69-4
104.149
GTCCGKPBSJZVRZ-UHFFFAOYSA-N
12262
CC(CC(C)O)O
C5H12O2
MFCD00004549
2,4-amylene glycol, 2,4-amyleneglycol, 2,4-dihydroxypentane, 2,4-pentandiol, 2,4-pentanediol, 2,4-pentanediol dl+meso, dl-2,4-pentanediol, isoamylene alcohol, pentanediol-2,4, wln: qy&1yq
pentane-2,4-diol

Specifications

0.955
Odorless
Hygroscopic
10g
C5H12O2
969186
Fully miscible.
CC(CC(C)O)O
104.149
104.15
2,4-Pentanediol, (+/-) + meso
101°C (213°F)
1.435
200°C to 201°C
625-69-4
MFCD00004549
2,4-amylene glycol, 2,4-amyleneglycol, 2,4-dihydroxypentane, 2,4-pentandiol, 2,4-pentanediol, 2,4-pentanediol dl+meso, dl-2,4-pentanediol, isoamylene alcohol, pentanediol-2,4, wln: qy&1yq
GTCCGKPBSJZVRZ-UHFFFAOYSA-N
pentane-2,4-diol
12262
99%
Safety and Handling

Safety and Handling

EINECSNumber : 210-907-5

RTECSNumber : SA0490000

TSCA : No

Recommended Storage : Ambient temperatures