missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Pentanediol, (+/-) + meso, 99%

Manufacturer:  Alfa Aesar™ L0177918

 View more versions of this product

Catalog No. AAL0177918



Description

Description

Applications
2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.

Solubility
Fully miscible.

Notes
Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

0.955
Odorless
Hygroscopic
50g
625-69-4
MFCD00004549
2,4-amylene glycol, 2,4-amyleneglycol, 2,4-dihydroxypentane, 2,4-pentandiol, 2,4-pentanediol, 2,4-pentanediol dl+meso, dl-2,4-pentanediol, isoamylene alcohol, pentanediol-2,4, wln: qy&1yq
GTCCGKPBSJZVRZ-UHFFFAOYSA-N
pentane-2,4-diol
12262
99%
101°C (213°F)
1.435
200°C to 201°C
(+/-) + meso
C5H12O2
969186
Fully miscible.
CC(CC(C)O)O
104.149
104.15

Chemical Identifiers

625-69-4
104.149
GTCCGKPBSJZVRZ-UHFFFAOYSA-N
12262
CC(CC(C)O)O
C5H12O2
MFCD00004549
2,4-amylene glycol, 2,4-amyleneglycol, 2,4-dihydroxypentane, 2,4-pentandiol, 2,4-pentanediol, 2,4-pentanediol dl+meso, dl-2,4-pentanediol, isoamylene alcohol, pentanediol-2,4, wln: qy&1yq
pentane-2,4-diol
Safety and Handling

Safety and Handling

EINECSNumber : 210-907-5

RTECSNumber : SA0490000

TSCA : No

Recommended Storage : Ambient temperatures

Documents