missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Octanedione, 98+%

$133.54 - $2,021.72

Chemical Identifiers

CAS 14090-87-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00015259
InChI Key GJYXGIIWJFZCLN-UHFFFAOYSA-N
Synonym 2,4-octanedione, 2,4-octanedion, acmc-1bu2j
PubChem CID 84192
IUPAC Name octane-2,4-dione
SMILES CCCCC(=O)CC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1016109 View Documents Alfa Aesar™
A1016109
10g Each for $133.54
 
AAA1016118 View Documents Alfa Aesar™
A1016118
50g Each for $507.35
 
AAA1016130 View Documents Alfa Aesar™
A1016130
250g Each for $2,021.72
 
Specifications

Chemical Identifiers

14090-87-0
142.198
GJYXGIIWJFZCLN-UHFFFAOYSA-N
84192
CCCCC(=O)CC(=O)C
C8H14O2
MFCD00015259
2,4-octanedione, 2,4-octanedion, acmc-1bu2j
octane-2,4-dione

Specifications

71°C (159°F)
10g
C8H14O2
1751147
GJYXGIIWJFZCLN-UHFFFAOYSA-N
octane-2,4-dione
84192
≥98%
97°C to 99°C (27mmHg)
14090-87-0
MFCD00015259
2,4-octanedione, 2,4-octanedion, acmc-1bu2j
CCCCC(=O)CC(=O)C
142.198
142.2
2,4-Octanedione